o-Xylylenebis(triphenylphosphonium bromide) ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(1,2-Phenylenebis(methylene))bis(triphenylphosphonium) bromide
L03103.09
L03103.18
L03103.09EA
0
EUR
L03103.09
L03103.18
o-Xylylenebis(triphenylphosphonium bromide) ≥98%
o-Xylylenebis(triphenylphosphonium bromide)
Purity: 98+%
CAS No.: 1519-46-6
Molecular Formula: C44H38Br2P2
Formula Weight: 788.55
Odour: Odourless
Melting Point:
Formula:
C₄₄H₃₈Br₂P₂ MW: 788,55 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044843 CAS Number: 1519-46-6 EINECS: 216-183-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...