(±)-3-Amino-4,4,4-trifluorobutyric acid hydrochloride 95%
Supplier: Thermo Scientific
|
Warning
|
L18336.03
L18336.03EA
227
EUR
L18336.03
(±)-3-Amino-4,4,4-trifluorobutyric acid hydrochloride 95%
(±)-3-Amino-4,4,4-trifluorobutyric acid hydrochloride
Purity: 95%
CAS No.: 91291-66-6
Molecular Formula: C4H6F3NO2.HCl
Formula Weight: 193.55
Melting Point: 197-199°
MDL No.: MFCD00181951
Formula:
C₄H₇ClF₃NO₂ MW: 193,55 g/mol Melting Pt: 197…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00181951 CAS Number: 91291-66-6 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...