(Methylthio)acetonitrile ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
L03066.06
L03066.14
L03066.22
L03066.06EA
34.5
EUR
L03066.06
L03066.14
L03066.22
(Methylthio)acetonitrile ≥99%
(Methylthio)acetonitrile
Purity: 99%
CAS No.: 35120-10-6
Molecular Formula: C3H5NS
Formula Weight: 87.14
Flash Point: 67°(152°F)
Refractive Index: 1.4820
Boiling Point: 166-167°
Beilstein Reference: 1738015
MDL No.: MFCD00001925
Formula:
C₃H₅NS Boiling Pt: 166…167 °C Density: 1,052 Flash Pt: 67 °C (152 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001925 CAS Number: 35120-10-6 EINECS: 252-383-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...