1-Acetyl-2-phenylhydrazine for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.20561.0050 8.20561.0010
8.20561.0050EA 126 EUR
8.20561.0050 8.20561.0010
1-Acetyl-2-phenylhydrazine for synthesis, Sigma-Aldrich®
1-Acetyl-2-phenylhydrazine
Formula: C₈H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD00008672
CAS Number: 114-83-0
EINECS: 204-055-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 131 °C
Molar Mass 150.18 g/mol

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR