N,N-Diethylhydroxylamine 97%

Supplier: Thermo Fisher Scientific
Warning

114421000 114425000
ACRO114421000EA 45.2 EUR
ACRO114421000 ACRO114425000
N,N-Diethylhydroxylamine 97%
N,N-Diethylhydroxylamine
Formula: C₄H₁₁NO
Melting Pt: 26…25 °C
Density: 0,867 g/cm³
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00002126
CAS Number: 3710-84-7
EINECS: 223-055-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4220 (20°C, 589 nm)
Water ≤1 %
GC ≥96.0 %
Color scale ≤100 APHA
Copper (Cu) ≤20 ppm
Iron (Fe) ≤400 ppm
Lead (Pb) ≤5 ppm
Mercury (Hg) ≤1 ppm
Zinc (Zn) ≤20 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.2 % (in HCl)
Loss on ignition ≤8 % (800°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR