(p-Toluenesulphonyl)methyl isocyanide 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1-((Isocyanomethyl)sulphonyl)-4-methylbenzene, Tosylmethyl isocyanide
182450250
182450050
182451000
ACRO182450250EA
154
EUR
ACRO182450250
ACRO182450050
ACRO182451000
(p-Toluenesulphonyl)methyl isocyanide 98%
(p-Toluenesulphonyl)methyl isocyanide
Formula:
C₉H₉NO₂S MW: 195,24 g/mol Melting Pt: 111…113 °C Storage Temperature: Freezer |
MDL Number:
MFCD00000005 CAS Number: 36635-61-7 EINECS: 253-140-1 UN: 2811 ADR: 6.1,II |
Specification Test Results
Appearance | Light yellow to beige to light brown Powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 109 to 115 °C (Dec) |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...