Ethyl-5-fluorobenzo[b]thiophene-2-carboxylate 96%
Supplier: Thermo Fisher Scientific
H34167.03
H34167.06
H34167.03EA
0
EUR
H34167.03
H34167.06
Ethyl-5-fluorobenzo[b]thiophene-2-carboxylate 96%
Ethyl 5-fluorobenzo[b]thiophene-2-carboxylate
Purity: 96%
CAS No.: 13771-69-2
Molecular Formula: C11H9FO2S
Formula Weight: 224.26
Melting Point: 88-89°
MDL No.: MFCD12923024
Formula:
C₁₁H₉FO₂S Melting Pt: 88…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD12923024 CAS Number: 13771-69-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...