2-Naphthonitrile ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Naphthalene-2-carbonitrile

L03704.06 L03704.14
L03704.06EA 50.9 EUR
L03704.06 L03704.14
2-Naphthonitrile ≥97%
2-Naphthonitrile

Purity: 97% CAS No.: 613-46-7 Molecular Formula: C11H7N Formula Weight: 153.18 Melting Point: 63-67° Boiling Point: 156-158°/12mm UN No.: 3439 MDL No.: MFCD00016807

Formula: C₁₁H₇N
Boiling Pt: 156…158 °C (12 mmHg)
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00016807
CAS Number: 613-46-7
EINECS: 210-344-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR