2-Nitrocinnamaldehyde predominantly trans 98%
|
Warning
|
Synonyms:
2'-Nitrocinnamaldehyde
A10308.14
A10308.14EA
72.2
EUR
A10308.14
2-Nitrocinnamaldehyde predominantly trans 98%
2-Nitrocinnamaldehyde
Purity: 98%
CAS No.: 1466-88-2
Molecular Formula: C9H7NO3
Formula Weight: 177.16
Odour: Characteristic
Melting Point: 125-129°
Beilstein Reference: 908376
MDL No.: MFCD00007188
Formula:
C₉H₇NO₃ MW: 177,16 g/mol Melting Pt: 125…129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007188 CAS Number: 1466-88-2 EINECS: 215-988-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...