1,3,2-Benzodioxaborole (catecholborane) ≥97%

Supplier: Thermo Scientific
Danger

L14998.06 L14998.14
L14998.06EA 85.8 EUR
L14998.06 L14998.14
1,3,2-Benzodioxaborole (catecholborane) ≥97%
1,3,2-Benzodioxaborole (Catecholborane)

Purity: 97% CAS No.: 274-07-7 Molecular Formula: C6H5BO2 Formula Weight: 119.92 Storage Temperature: 2°C to 8°C Flash Point: 2°(35°F) Melting Point: 12° Boiling Point: 50°/50mm Beilstein Reference: 972072 UN No.: 2924 MDL No.: MFCD00005846

Formula: C₆H₅BO₂
MW: 119,92 g/mol
Boiling Pt: 50 °C (50 mmHg)
Melting Pt: 12 °C
Density: 1,125
Flash Pt: 2 °C (35 °F)
MDL Number: MFCD00005846
CAS Number: 274-07-7
EINECS: 205-991-5
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR