2,2'-Ethylenebis(1,3-dithiane) ≥98%
Supplier: Thermo Scientific
L16255.03
L16255.06
L16255.03EA
0
EUR
L16255.03
L16255.06
2,2'-Ethylenebis(1,3-dithiane) ≥98%
2,2'-Ethylenebis(1,3-dithiane)
Purity: 98+%
CAS No.: 14947-53-6
Molecular Formula: C10H18S4
Formula Weight: 266.51
Melting Point: 132-135°
Beilstein Reference: 1237944
Formula:
C₁₀H₁₈S₄ MW: 266,51 g/mol Melting Pt: 132…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040467 CAS Number: 14947-53-6 EINECS: 239-022-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...