1-Chlorophthalazin-4-one ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
H27515.06
H27515.03
H27515.06EA
330
EUR
H27515.06
H27515.03
1-Chlorophthalazin-4-one ≥98%
1-Chlorophthalazin-4-one
Purity: 98%
CAS No.: 2257-69-4
Molecular Formula: C8H5ClN2O
Formula Weight: 180.59
Melting Point: 270-274°
MDL No.: MFCD00102192
Formula:
C₈H₅ClN₂O Melting Pt: 270…274 °C Storage Temperature: Ambient |
MDL Number:
MFCD00102192 CAS Number: 2257-69-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...