N-Ethylpropylamine ≥97%
Supplier: Thermo Scientific
|
Danger
|
14846.06
14846.14
14846.06EA
92.5
EUR
14846.06
14846.14
N-Ethylpropylamine ≥97%
N-Ethylpropylamine
Purity: 97+%
CAS No.: 20193-20-8
Molecular Formula: CH3CH2NHCH2CH2CH3
Formula Weight: 87.16
Flash Point: 4.4°(40°F)
Physical Form: Liquid
Density: 0.748
Boiling Point: 80-85°
UN No.: 2733
MDL No.: MFCD00015198
Formula:
C₅H₁₃N MW: 87,16 g/mol Boiling Pt: 80…85 °C Density: 0,748 Flash Pt: 4,4 °C (40 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015198 CAS Number: 20193-20-8 EINECS: 243-573-4 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...