Tripalmitin (glycerol tripalmitate) 99%

Supplier: Thermo Scientific
Warning

Synonyms: Propane-1,2,3-triyl tripalmitate, Tripalmitin, Glycerol tripalmitate

215670050 215670250 215670010
ACRO215670050EA 125 EUR
ACRO215670050 ACRO215670250 ACRO215670010
Tripalmitin (glycerol tripalmitate) 99%
Tripalmitin (Glycerol tripalmitate)
Formula: C₅₁H₉₈O₆
MW: 807,34 g/mol
Boiling Pt: 315 °C
Melting Pt: 65…68 °C
Storage Temperature: Freezer
MDL Number: MFCD00008995
CAS Number: 555-44-2
EINECS: 209-098-1

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR