Tripalmitin (glycerol tripalmitate) ≥98%

Supplier: Thermo Scientific

Synonyms: Propane-1,2,3-triyl tripalmitate, Tripalmitin, Glycerol tripalmitate

A10922.06 A10922.14 A10922.03
A10922.06EA 122 EUR
A10922.06 A10922.14 A10922.03
Tripalmitin (glycerol tripalmitate) ≥98%
Tripalmitin (Glycerol tripalmitate)

Purity: 98% CAS No.: 555-44-2 Molecular Formula: C51H98O6 Formula Weight: 807.34 Melting Point: 60-64° MDL No.: MFCD00008995

Formula: C₅₁H₉₈O₆
MW: 807,34 g/mol
Melting Pt: 60…64 °C
Storage Temperature: Freezer
MDL Number: MFCD00008995
CAS Number: 555-44-2
EINECS: 209-098-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR