2,6-Dibromo-4-fluorphenyl isocyanate tech. 90%, Technical Grade

Supplier: Thermo Fisher Scientific
Danger

L18744.03 L18744.06
L18744.03EA 0 EUR
L18744.03 L18744.06
2,6-Dibromo-4-fluorphenyl isocyanate tech. 90%, Technical Grade
2,6-Dibromo-4-fluorphenyl isocyanate

Purity: 90% CAS No.: 76393-18-5 Molecular Formula: C7H2Br2FNO Formula Weight: 294.91 Flash Point: 110°(230°F) Melting Point: 42-45° Beilstein Reference: 8617287 UN No.: 2811 MDL No.: MFCD00037041

Formula: C₇H₂Br₂FNO
Melting Pt: 42…45 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00037041
CAS Number: 76393-18-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR