4-Octyloxybenzonitrile 98
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
4-n-Octyloxybenzonitrile
A12843.06
A12843.14
A12843.22
A12843.06EA
37.5
EUR
A12843.06
A12843.14
A12843.22
4-Octyloxybenzonitrile 98
4-Octyloxybenzonitrile
Purity: 98%
CAS No.: 88374-55-4
Molecular Formula: C15H21NO
Formula Weight: 231.34
Refractive Index: 1.5100
Melting Point: 24-26°
Boiling Point: 171-173°/2mm
Beilstein Reference: 2844884
UN No.: 3439
MDL No.: MFCD00043483
Formula:
C₁₅H₂₁NO MW: 231.34 g/mol Boiling Pt: 171…173 °C (2 mmHg) Melting Pt: 24…26 °C Density: 0.937 Storage Temperature: Ambient |
MDL Number:
MFCD00043483 CAS Number: 88374-55-4 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...