Bis(pentamethylcyclopentadienyl)magnesium ≥99.999% (metals basis)
Supplier: Thermo Scientific
|
Danger
|
38598.04
38598.09
38598.04EA
0
EUR
38598.04
38598.09
Bis(pentamethylcyclopentadienyl)magnesium ≥99.999% (metals basis)
Bis(pentamethylcyclopentadienyl)magnesium
Purity: 99.999%
CAS No.: 74507-64-5
Molecular Formula: C20H30Mg
Formula Weight: 294.77
Physical Form: Crystalline, Packaged in ampoules
Sensitivity: Air and Moisture Sensitive
UN No.: 3181
MDL No.: MFCD00151387
Formula:
C₂₀H₃₀Mg MW: 294,757 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00151387 CAS Number: 74507-64-5 UN: 3181 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...