(Pentamethylcyclopentadienyl)zirconium trichloride ≥99%
Supplier: Thermo Scientific
|
Warning
|
39551.03
39551.06
39551.03EA
138
EUR
39551.03
39551.06
(Pentamethylcyclopentadienyl)zirconium trichloride ≥99%
(Pentamethylcyclopentadienyl)zirconium trichloride
Purity: 99%
CAS No.: 75181-07-6
Molecular Formula: C10H15Cl3Zr
Formula Weight: 332.81
Physical Form: Powder. Packaged in ampoules
Sensitivity: Moisture Sensitive
Appearance: Yellow to yellow-green
Melting Point: 230° dec.
MDL No.: MFCD00070444
Formula:
C₁₀H₁₅Cl₃Zr MW: 332,81 g/mol Melting Pt: 230 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00070444 CAS Number: 75181-07-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...