Benzyltrimethylammonium dichloroiodate 99%

Supplier: Thermo Fisher Scientific
Warning

268600250 268600050 268601000
ACRO268600250EA 180 EUR
ACRO268600250 ACRO268600050 ACRO268601000
Benzyltrimethylammonium dichloroiodate 99%
Benzyltrimethylammonium dichloroiodate
Formula: C₁₀H₁₆Cl₂IN
Melting Pt: 125…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00075259
CAS Number: 114971-52-7

Order Now

Specification Test Results

Appearance Yellow to light brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 125 to 128 °C
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR