Benzyltrimethylammonium dichloroiodate 95%
Supplier: Thermo Scientific
|
Warning
|
B22892.06
B22892.14
B22892.06EA
61.3
EUR
B22892.06
B22892.14
Benzyltrimethylammonium dichloroiodate 95%
Benzyltrimethylammonium dichloroiodate
Purity: 95%
CAS No.: 114971-52-7
Molecular Formula: C10H16NICl2
Formula Weight: 348.06
Melting Point: ca. 125°
Beilstein Reference: 4611949
Formula:
C₁₀H₁₆Cl₂IN MW: 348,06 g/mol Melting Pt: ∼, 125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075259 CAS Number: 114971-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...