7-Iodooxindole 96%

Supplier: Thermo Fisher Scientific

H52337.MD H52337.03
H52337.MDEA 0 EUR
H52337.MD H52337.03
7-Iodooxindole 96%
7-Iodooxindole

Purity: 96% CAS No.: 31676-49-0 Molecular Formula: C8H6INO Formula Weight: 259.04 Melting Point: 208-209° MDL No.: MFCD09835634

Formula: C₈H₆INO
MW: 259.04 g/mol
Melting Pt: 208…209 °C
Storage Temperature: Ambient
MDL Number: MFCD09835634
CAS Number: 31676-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR