2-Cyanopyrazine ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Pyrazine-2-carbonitrile, 2-Pyrazinecarbonitril, Pyrazinecarbonitrile
A10974.06
A10974.14
A10974.22
A10974.06EA
27.6
EUR
A10974.06
A10974.14
A10974.22
2-Cyanopyrazine ≥97%
2-Cyanopyrazine
Purity: 97+%
CAS No.: 19847-12-2
Molecular Formula: C5H3N3
Formula Weight: 105.10
Flash Point: 96°(204°F)
Refractive Index: 1.5360
Melting Point: 18-20°
Boiling Point: 90-93°/15mm
Beilstein Reference: 110008
UN No.: 3276
MDL No.: MFCD00049361
Formula:
C₅H₃N₃ MW: 105.10 g/mol Boiling Pt: 90…93 °C (15 mmHg) Melting Pt: 18…20 °C Density: 1.174 Flash Pt: 96 °C (204 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00049361 CAS Number: 19847-12-2 EINECS: 243-369-5 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...