Octyltrimethylammonium bromide ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Trimethyl-1-octylammonium bromide
L08610.06
L08610.14
L08610.22
L08610.06EA
39.2
EUR
L08610.06
L08610.14
L08610.22
Octyltrimethylammonium bromide ≥97%
Octyltrimethylammonium bromide
Purity: 97%
CAS No.: 2083-68-3
Molecular Formula: CH3(CH2)7N(CH3)3Br
Formula Weight: 252.24
Melting Point: 220-222°
Beilstein Reference: 3628448
Formula:
C₁₁H₂₆BrN Melting Pt: 220…222 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051873 CAS Number: 2083-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...