3-Bromo-4-pyridone ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Bromo-4-hydroxypyridine, 3-Bromo-4-pyridinol

H33754.03 H33754.06
H33754.03EA 0 EUR
H33754.03 H33754.06
3-Bromo-4-pyridone ≥97%
3-Bromo-4-pyridone

Purity: 97% CAS No.: 36953-41-0 Molecular Formula: C5H4BrNO Formula Weight: 174.00 Melting Point: 225-230° MDL No.: MFCD01646107

Formula: C₅H₄BrNO
MW: 174,00 g/mol
Melting Pt: 225…230 °C
Storage Temperature: Ambient
MDL Number: MFCD01646107
CAS Number: 36953-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR