Aminoiminomethanesulphinic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Thiourea dioxide, FormamidinesulphInic acid
M-Clarity™ quality level = MQ200
8.20082.1000
8.20082.0250
8.20082.1000EA
157
EUR
8.20082.1000
8.20082.0250
Aminoiminomethanesulphinic acid for synthesis, Sigma-Aldrich®
Aminoiminomethanesulphinic acid
Formula:
CH₄N₂O₂S MW: 108.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002397 CAS Number: 1758-73-2 EINECS: 217-157-8 UN: 3341 ADR: 4.2 II |
Specification Test Results
Solubility | 27 g/l (20 °C) |
Melting Point | 123 °C (decomposition) |
Molar Mass | 108.12 g/mol |
Bulk Density | 850 kg/m3 |
Density | 1.68 g/cm3 (20 °C) |
pH | 4 (10 g/l, H2O, 20 °C) |
Hazard Class | 4.2 |
Hazard Class Description | Spontaneously Combustible Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...