Mucochloric acid 99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2,3-Dichloromalealdehydic acid
128065000
128061000
ACRO128065000EA
109
EUR
ACRO128065000
ACRO128061000
Mucochloric acid 99%
Mucochloric acid
Formula:
C₄H₂Cl₂O₃ MW: 168,96 g/mol Melting Pt: 125…128 °C Flash Pt: 186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00135465 CAS Number: 87-56-9 EINECS: 201-752-4 UN: 2923 ADR: 8,II |
Specification Test Results
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 125 to 128 °C |
Water | ≤4 % (K.F.) |
Titration with NaOH | ≥98.5 % (on dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...