2-Amino-5-chlorobenzamide ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A11153.03
A11153.06
A11153.03EA
22.4
EUR
A11153.03
A11153.06
2-Amino-5-chlorobenzamide ≥98%
2-Amino-5-chlorobenzamide
Purity: 98+%
CAS No.: 5202-85-7
Molecular Formula: C7H7ClN2O
Formula Weight: 170.60
Melting Point: 169-173°
Beilstein Reference: 743347
Formula:
C₇H₇ClN₂O MW: 170.60 g/mol Melting Pt: 169…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017126 CAS Number: 5202-85-7 EINECS: 225-990-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...