Diethyl chlorophosphite ≥97%

Supplier: Thermo Fisher Scientific
Danger

L09919.06 L09919.14
L09919.06EA 63.4 EUR
L09919.06 L09919.14
Diethyl chlorophosphite ≥97%
Diethyl chlorophosphite

Purity: 97% CAS No.: 589-57-1 Molecular Formula: C4H10ClO2P Formula Weight: 156.55 Flash Point: 25°(77°F) Refractive Index: 1.4375 Odour: Acrid Boiling Point: 153-155° Beilstein Reference: 1098392 Merck Reference: 14,3841 UN No.: 2920 MDL No.: MFCD00009074

Formula: C₄H₁₀ClO₂P
MW: 156.55 g/mol
Boiling Pt: 153…155 °C
Density: 1.082
Flash Pt: 25 °C (77 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009074
CAS Number: 589-57-1
EINECS: 209-652-2
UN: 2920
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR