Diiron boride ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: Iron boride

22952.14 22952.22
22952.14EA 168 EUR
22952.14 22952.22
Diiron boride ≥98%
Diiron boride

Purity: 98% CAS No.: 12006-85-8 Molecular Formula: Fe2B Formula Weight: 122.50 Physical Form: -35 Mesh Odour: Odourless MDL No.: MFCD00016092

Formula: BFe₂
Storage Temperature: Ambient
MDL Number: MFCD00016092
CAS Number: 12006-85-8
EINECS: 234-490-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR