4-(2-Pyridylazo)resorcinol 97+%

Supplier: Thermo Scientific
Warning

Synonyms: 4-(2'-Pyridylazo)resorcinol

418590010 418590050 418590250
ACRO418590010EA 34.5 EUR
ACRO418590010 ACRO418590050 ACRO418590250
4-(2-Pyridylazo)resorcinol 97+%
4-(2-Pyridylazo)resorcinol
Formula: C₁₁H₉N₃O₂
MW: 215,21 g/mol
Melting Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00044237
CAS Number: 1141-59-9
EINECS: 214-528-6

Order Now

Specification Test Results

Appearance Orange Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR