2-Acetyl-5-cyanothiophene ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
5-Acetylthiophene-2-carbonitrile
A12765.02
A12765.88
A12765.02EA
39.6
EUR
A12765.02
A12765.88
2-Acetyl-5-cyanothiophene ≥97%
2-Acetyl-5-cyanothiophene
Purity: 97%
CAS No.: 88653-55-8
Molecular Formula: C7H5NOS
Formula Weight: 151.19
Melting Point: 76-80°
Boiling Point: 95-97°/0.5mm
UN No.: 3439
MDL No.: MFCD00052314
Formula:
C₇H₅NOS MW: 151,19 g/mol Boiling Pt: 95…97 °C (0,5 mmHg) Melting Pt: 76…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052314 CAS Number: 88653-55-8 UN: 3439 ADR: 6.1,III |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...