2-Acetyl-3-aminothiophene 97%

Supplier: Thermo Scientific
Danger

L10563.03 L10563.MD
L10563.03EA 127 EUR
L10563.03 L10563.MD
2-Acetyl-3-aminothiophene 97%
2-Acetyl-3-aminothiophene

Purity: 97% CAS No.: 31968-33-9 Molecular Formula: C6H7NOS Formula Weight: 141.19 Melting Point: 84-88° Boiling Point: 120-125°/0.8mm MDL No.: MFCD00052585

Formula: C₆H₇NOS
MW: 141,19 g/mol
Boiling Pt: 120…125 °C (0,8 mmHg)
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00052585
CAS Number: 31968-33-9
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR