α,α-Diphenyl-N-methyl-D-prolinol ≥97%, ee 99%
Supplier: Thermo Scientific
L19398.03
L19398.MD
L19398.03EA
253
EUR
L19398.03
L19398.MD
α,α-Diphenyl-N-methyl-D-prolinol ≥97%, ee 99%
α,α-Diphenyl-N-methyl-D-prolinol
Purity: 97%
CAS No.: 144119-12-0
Molecular Formula: C18H21NO
Formula Weight: 267.37
Storage Temperature: 2°C to 8°C
Melting Point: 68-71°
Beilstein Reference: 1535407
Optical Rotation: -52° (c=1 in chloroform)
MDL No.: MFCD00145217
Formula:
C₁₈H₂₁NO Melting Pt: 68…71 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00145217 CAS Number: 144119-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...