(R,R)-(+)-2,3-Bis(diphenylphosphino)butane ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(2R,3R)-Butane-2,3-diylbis(diphenylphosphine), (2R,3R)-(+)-Bis(diphenylphosphino)butane
H27448.MF
H27448.MD
H27448.03
H27448.MFEA
78.2
EUR
H27448.MF
H27448.MD
H27448.03
(R,R)-(+)-2,3-Bis(diphenylphosphino)butane ≥98%
(R,R)-(+)-2,3-Bis(diphenylphosphino)butane
Purity: 98%
CAS No.: 74839-84-2
Molecular Formula: C28H28P2
Formula Weight: 426.48
Melting Point: 104-109°
MDL No.: MFCD00075403
Formula:
C₂₈H₂₈P₂ MW: 426,48 g/mol Melting Pt: 104…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075403 CAS Number: 74839-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...