(S,S)-(-)-2,3-Bis(diphenylphosphino)butane ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: (2S,3S)-(-)-Bis(diphenylphosphino)butane, (2S,3S)-Butane-2,3-diylbis(diphenylphosphine), (S,S)-CHIRAPHOS

H27586.MD H27586.03
H27586.MDEA 190 EUR
H27586.MD H27586.03
(S,S)-(-)-2,3-Bis(diphenylphosphino)butane ≥98%
(S,S)-(-)-2,3-Bis(diphenylphosphino)butane

Purity: 98% CAS No.: 64896-28-2 Molecular Formula: C28H28P2 Formula Weight: 426.48 Melting Point: 107-109° Beilstein Reference: 4268869

Formula: C₂₈H₂₈P₂
MW: 426,47 g/mol
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00009836
CAS Number: 64896-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR