Bromomethyl acetate 95%

Supplier: Thermo Scientific
Warning

H56755.06 H56755.09
H56755.06EA 319 EUR
H56755.06 H56755.09
Bromomethyl acetate 95%
Bromomethyl acetate

Purity: 95% CAS No.: 590-97-6 Molecular Formula: C3H5BrO2 Formula Weight: 152.98 Storage Temperature: 2°C to 8°C Flash Point: 57°(135°F) Boiling Point: 130-133° UN No.: 3272 MDL No.: MFCD00000170

Formula: H₃CCOOCH₂Br
MW: 152,98 g/mol
Boiling Pt: 130…133 °C
Density: 1,56
Flash Pt: 57 °C (135 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000170
CAS Number: 590-97-6
EINECS: 209-696-2
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR