2-Naphthalenesulphonic acid ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Naphthalene-2-sulphonic acid, β-Naphthalenesulphonic acid
H56685.22
H56685.36
H56685.22EA
121
EUR
H56685.22
H56685.36
2-Naphthalenesulphonic acid ≥98%
2-Naphthalenesulphonic acid
Formula:
C₁₀H₈O₃S MW: 208,24 g/mol Boiling Pt: ∼350 °C (1013 hPa) Melting Pt: 124 °C Density: 1,441 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149271 CAS Number: 120-18-3 EINECS: 204-375-3 UN: 2585 ADR: 8,III Merck Index: 12,06464 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...