4-Methylphenylacetone ≥97%
Supplier: Thermo Fisher Scientific
B24728.03
B24728.06
B24728.03EA
34.4
EUR
B24728.03
B24728.06
4-Methylphenylacetone ≥97%
4-Methylphenylacetone
Purity: 98%
CAS No.: 2096-86-8
Molecular Formula: C10H12O
Formula Weight: 148.20
MDL No.: MFCD00017250
Formula:
C₁₀H₁₂O Storage Temperature: Ambient |
MDL Number:
MFCD00017250 CAS Number: 2096-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...