4-Cyanophenylacetonitrile ≥97%

Supplier: Thermo Fisher Scientific
Danger

H54112.06 H54112.03
H54112.06EA 201 EUR
H54112.06 H54112.03
4-Cyanophenylacetonitrile ≥97%
4-Cyanophenylacetonitrile

Purity: 97% CAS No.: 876-31-3 Molecular Formula: C9H6N2 Formula Weight: 142.16 Flash Point: 71°(160°F) Refractive Index: 1.499 Boiling Point: 180-181° MDL No.: MFCD03839848

Formula: C₉H₆N₂
Boiling Pt: 180…181 °C
Density: 1,056
Flash Pt: 71 °C (160 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060305
CAS Number: 876-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR