2-Coumaranone 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Benzofuran-2(3H)-one, Benzo[b]furan-2(3H)-one
154460010
154460050
154460100
ACRO154460010EA
18
EUR
ACRO154460010
ACRO154460050
ACRO154460100
2-Coumaranone 97%
2-Coumaranone
Formula:
C₈H₆O₂ MW: 134,13 g/mol Melting Pt: 47…52 °C Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005856 CAS Number: 553-86-6 EINECS: 209-052-0 |
Specification Test Results
Appearance | White to yellow to light orange Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 47 to 52 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...