4-Chlorophenyl phosphorodichloridate 98+%, rods

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Chlorophenyl dichlorophosphate

A15507.06 A15507.14 A15507.22
A15507.06EA 27.2 EUR
A15507.06 A15507.14 A15507.22
4-Chlorophenyl phosphorodichloridate 98+%, rods
4-Chlorophenyl phosphorodichloridate

Purity: 98+% CAS No.: 772-79-2 Molecular Formula: C6H4Cl3O2P Formula Weight: 245.43 Flash Point: 106°(222°F) Refractive Index: 1.5398 Odour: Acrid Boiling Point: 113-115°/0.9mm Beilstein Reference: 910754 UN No.: 3265 MDL No.: MFCD00009705

Formula: C₆H₄Cl₃O₂P
MW: 245.43 g/mol
Boiling Pt: 113…115 °C (0.9 mmHg)
Density: 1.508
Flash Pt: 106 °C (222 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009705
CAS Number: 772-79-2
EINECS: 212-254-1
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR