Trimethyl phosphonoacetate ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Methyl 2-(dimethoxyphosphoryl)acetate, Dimethyl methoxycarbonylmethanephosphonate
A13301.14
A13301.22
A13301.36
A13301.14EA
49.5
EUR
A13301.14
A13301.22
A13301.36
Trimethyl phosphonoacetate ≥98%
Trimethyl phosphonoacetate
Purity: 98%
CAS No.: 5927-18-4
Molecular Formula: C5H11O5P
Formula Weight: 182.11
Flash Point: 70°(158°F)
Refractive Index: 1.4370
Boiling Point: 119-120°/1mm
Beilstein Reference: 1865177
MDL No.: MFCD00008452
Formula:
C₅H₁₁O₅P MW: 182,11 g/mol Boiling Pt: 119…120 °C (1 mmHg) Density: 1,263 Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008452 CAS Number: 5927-18-4 EINECS: 227-663-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...