Piperonylacetone ≥98%

Supplier: Thermo Scientific

Synonyms: 4-(3,4-Methylenedioxy)phenyl-2-butanone

L06258.06 L06258.14
L06258.06EA 38.5 EUR
L06258.06 L06258.14
Piperonylacetone ≥98%
Piperonylacetone

Purity: 98% CAS No.: 55418-52-5 Molecular Formula: C11H12O3 Formula Weight: 192.22 Flash Point: 110°(230°F) Melting Point: 49-53° Boiling Point: 164-165°/12mm Beilstein Reference: 169843 MDL No.: MFCD00016910

Formula: C₁₁H₁₂O₃
MW: 192,22 g/mol
Boiling Pt: 164…165 °C (12 mmHg)
Melting Pt: 49…53 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00016910
CAS Number: 55418-52-5
EINECS: 259-630-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR