Chloroform-[¹³C] (99% ¹³C )

Supplier: LGC Standards PROMOCHEM
Warning

CILCLM-262-0.1 CIL-CLM-262-1
PROOCILCLM-262-0.1EA 356 EUR
PROOCILCLM-262-0.1 PROOCIL-CLM-262-1
Chloroform-[¹³C] (99% ¹³C )
Chloroform-[13C]
Formula: CHCl₃
Storage Temperature: Ambient
CAS Number: 31717-44-9
UN: 1888
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR