3-Formylfuran-2-boronic acid ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (3-Formylfuran-2-yl)boronic acid

L15198.03 L15198.06
L15198.03EA 0 EUR
L15198.03 L15198.06
3-Formylfuran-2-boronic acid ≥97%
3-Formylfuran-2-boronic acid

Purity: 97% CAS No.: 27339-38-4 Molecular Formula: C5H5BO4 Formula Weight: 139.90 Melting Point: ca 150° dec. Beilstein Reference: 1637137

Formula: C₅H₅BO₄
MW: 139.90 g/mol
Melting Pt: ∼ 150 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01075680
CAS Number: 27339-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR