5-Formyl-2-furanylboronic acid ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
L17920.03
L17920.06
L17920.03EA
42.7
EUR
L17920.03
L17920.06
5-Formyl-2-furanylboronic acid ≥97%
5-Formyl-2-furanylboronic acid
Purity: 97%
CAS No.: 27329-70-0
Molecular Formula: C5H5BO4
Formula Weight: 139.90
Melting Point: ca 140° dec.
Beilstein Reference: 1637050
Formula:
C₅H₅BO₄ Melting Pt: ∼ 140 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD01114696 CAS Number: 27329-70-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...