1-(2-Thiazolylazo)-2-naphthol ≥98%

Supplier: Thermo Fisher Scientific
Warning

B24011.03 B24011.06 B24011.14
B24011.03EA 48.7 EUR
B24011.03 B24011.06 B24011.14
1-(2-Thiazolylazo)-2-naphthol ≥98%
1-(2-Thiazolylazo)-2-naphthol

Purity: 98% CAS No.: 1147-56-4 Molecular Formula: C13H9N3OS Formula Weight: 255.30 Melting Point: 138-140° Beilstein Reference: 223783

Formula: C₁₃H₉N₃OS
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00021611
CAS Number: 1147-56-4
EINECS: 214-555-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR