Ethyl-3-(trifluoromethyl)crotonate mixture E/Z 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-(Trifluoromethyl)crotonic acid ethyl ester
L09688.09
L09688.04
L09688.09EA
270
EUR
L09688.09
L09688.04
Ethyl-3-(trifluoromethyl)crotonate mixture E/Z 96%
Ethyl-3-(trifluoromethyl)crotonate
Purity: 96%
CAS No.: 24490-03-7
Molecular Formula: C7H9F3O2
Formula Weight: 182.14
Flash Point: 32°(89°F)
Refractive Index: 1.3780
Boiling Point: 129-130°
Beilstein Reference: 1910081
UN No.: 3272
MDL No.: MFCD00040846
Formula:
C₇H₉F₃O₂ MW: 182,14 g/mol Boiling Pt: 129…130 °C Flash Pt: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040846 CAS Number: 24490-03-7 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...