Ethyl-3-amino-4,4,4-trifluorocrotonate 97%

Supplier: Thermo Scientific
Danger

Synonyms: Ethyl 3-amino-4,4,4-trifluorobut-2-enoate

B22545.06 B22545.14
B22545.06EA 41.5 EUR
B22545.06 B22545.14
Ethyl-3-amino-4,4,4-trifluorocrotonate 97%
Ethyl 3-amino-4,4,4-trifluorocrotonate

Purity: 97% CAS No.: 372-29-2 Molecular Formula: C6H8F3NO2 Formula Weight: 183.13 Flash Point: 65°(149°F) Refractive Index: 1.4229 Odour: Amine-like Melting Point: 24-26° Boiling Point: 83-85°/15mm Beilstein Reference: 4397839 UN No.: 3259 MDL No.: MFCD00068195

Formula: C₆H₈F₃NO₂
MW: 183,13 g/mol
Boiling Pt: 83…85 °C (15 mmHg)
Melting Pt: 24…26 °C
Density: 1,245
Flash Pt: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00068195
CAS Number: 372-29-2
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR